Assay Details
| Gene Symbol: Cyp27b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053763.1 | NP_446215.1 | 7 - 8 | 1216 | 108 | |
| XM_008765374.1 | XP_008763596.1 | 8 - 9 | 3088 | 108 | ||
| GenBank mRNA | AB001992.1 | - | 7 - 8 | 1216 | 108 | |
| AF000139.1 | - | 7 - 8 | 1208 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp27b1
Genomic Map