Assay Details
| Gene Symbol: Cuzd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054005.1 | NP_446457.1 | 3 - 4 | 701 | 84 | |
| GenBank mRNA | AF022147.1 | - | 3 - 4 | 701 | 84 | |
| AF167170.1 | - | 3 - 4 | 467 | 84 | ||
| BC090345.1 | - | 3 - 4 | 447 | 84 | ||
| BC107927.1 | - | 502 | 84 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Cuzd1
Genomic Map