Assay Details
| Gene Symbol: Park7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277249.1 | NP_001264178.1 | 2 - 3 | 209 | 117 | |
| NM_001277250.1 | NP_001264179.1 | 2 - 3 | 209 | 117 | ||
| NM_001277251.1 | NP_001264180.1 | 3 - 4 | 268 | 117 | ||
| NM_001277252.1 | NP_001264181.1 | 2 - 3 | 149 | 117 | ||
| NM_001277253.1 | NP_001264182.1 | 2 - 3 | 261 | 117 | ||
| NM_057143.2 | NP_476484.1 | 2 - 3 | 391 | 117 | ||
| GenBank mRNA | AF157511.1 | - | 2 - 3 | 391 | 117 | |
| AF157512.1 | - | 2 - 3 | 145 | 117 | ||
| AJ007291.1 | - | 3 - 4 | 316 | 117 | ||
| BC091128.1 | - | 2 - 3 | 182 | 7376914 | 117 | |
| FM055257.1 | - | 2 - 3 | 179 | 117 | ||
| FM061417.1 | - | 2 - 3 | 130 | 117 | ||
| FM069664.1 | - | 2 - 3 | 200 | 117 | ||
| FQ220895.1 | - | 2 - 3 | 261 | 117 | ||
| FQ223564.1 | - | 2 - 3 | 259 | 117 | ||
| FQ224480.1 | - | 3 - 4 | 252 | 117 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Park7
Genomic Map