Assay Details
| Gene Symbol: Klhl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057191.1 | NP_476539.1 | 2 - 3 | 1268 | 125 | |
| GenBank mRNA | AJ293948.1 | - | 2 - 3 | 1268 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klhl41
Genomic Map