Assay Details
| Gene Symbol: Daxx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080891.2 | NP_543167.2 | 2 - 3 | 297 | 78 | |
| XM_006256103.2 | XP_006256165.1 | 2 - 3 | 1079 | 78 | ||
| XM_006256104.2 | XP_006256166.1 | 2 - 3 | 753 | 78 | ||
| XM_006256105.2 | XP_006256167.1 | 2 - 3 | 649 | 78 | ||
| XM_006256106.2 | XP_006256168.1 | 2 - 3 | 315 | 78 | ||
| XM_006256107.2 | XP_006256169.1 | 2 - 3 | 419 | 78 | ||
| XM_006256108.2 | XP_006256170.1 | 2 - 3 | 299 | 78 | ||
| XM_006256109.1 | XP_006256171.1 | 1 - 2 | 1172 | 78 | ||
| XM_006256110.2 | XP_006256172.1 | 2 - 3 | 298 | 78 | ||
| XM_008772704.1 | XP_008770926.1 | 1 - 2 | 1172 | 78 | ||
| GenBank mRNA | AB064671.1 | - | 1 - 2 | 201 | 78 | |
| BC128729.1 | - | 2 - 3 | 297 | 8366304 | 78 | |
| FQ223571.1 | - | 2 - 3 | 268 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Daxx
Genomic Map