Assay Details
| Gene Symbol: Bpnt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171990.2 | NP_741987.1 | 5 - 6 | 449 | 76 | |
| XM_006250429.2 | XP_006250491.1 | 5 - 6 | 491 | 76 | ||
| XM_006250430.2 | XP_006250492.1 | 5 - 6 | 546 | 76 | ||
| XM_006250431.2 | XP_006250493.1 | 5 - 6 | 545 | 76 | ||
| GenBank mRNA | AJ000347.1 | - | 5 - 6 | 400 | 76 | |
| BC085692.1 | - | 5 - 6 | 449 | 7190529 | 76 | |
| FQ214355.1 | - | 5 - 6 | 472 | 76 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Bpnt1
Genomic Map