Assay Details
| Gene Symbol: Coq7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012785.3 | NP_036917.2 | 2 - 3 | 309 | 97 | |
| XM_006230108.2 | XP_006230170.1 | 2 - 3 | 383 | 97 | ||
| GenBank mRNA | FQ225865.1 | - | 309 | 97 | ||
| U46149.1 | - | 1 - 2 | 135 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Coq7
Genomic Map