Assay Details
| Gene Symbol: Proc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012803.1 | NP_036935.1 | 5 - 6 | 444 | 73 | |
| XM_006254526.2 | XP_006254588.1 | 5 - 6 | 516 | 73 | ||
| XM_006254527.2 | XP_006254589.1 | 5 - 6 | 523 | 73 | ||
| XM_008772018.1 | XP_008770240.1 | 5 - 6 | 586 | 73 | ||
| GenBank mRNA | BC078879.1 | - | 4 - 5 | 421 | 7122006 | 73 |
| X64336.1 | - | 5 - 6 | 444 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Proc
Genomic Map