Assay Details
| Gene Symbol: LOC103690160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230612.2 | XP_006230674.1 | 7 - 8 | 6770 | 70 | |
| XM_008760041.1 | XP_008758263.1 | 6 - 7 | 6749 | 70 | ||
| XM_008760042.1 | XP_008758264.1 | 5 - 6 | 6740 | 70 | ||
| Gene Symbol: Shank2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004133.1 | NP_001004133.1 | 8 - 9 | 1201 | 70 | |
| NM_133440.1 | NP_597684.1 | 9 - 10 | 1328 | 70 | ||
| NM_133441.1 | NP_597685.1 | 14 - 15 | 1822 | 70 | ||
| NM_201350.1 | NP_958738.1 | 21 - 22 | 2641 | 70 | ||
| XM_008760068.1 | XP_008758290.1 | 22 - 23 | 2582 | 70 | ||
| GenBank mRNA | AF060116.1 | - | 8 - 9 | 1201 | 70 | |
| AJ131899.1 | - | 9 - 10 | 1328 | 70 | ||
| AJ249562.1 | - | 14 - 15 | 1822 | 70 | ||
| AY298755.1 | - | 21 - 22 | 2641 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103690160
Genomic Map