Assay Details
| Gene Symbol: Atp12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301664.1 | NP_001288593.1 | 4 - 5 | 956 | 86 | |
| NM_133517.2 | NP_598201.2 | 5 - 6 | 815 | 86 | ||
| GenBank mRNA | M90398.1 | - | 5 - 6 | 739 | 86 | |
| U94912.1 | - | 5 - 6 | 811 | 86 | ||
| U94913.1 | - | 4 - 5 | 955 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp12a
Genomic Map