Assay Details
| Gene Symbol: Gbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133624.2 | NP_598308.1 | 8 - 9 | 1541 | 137 | |
| GenBank mRNA | BC086991.1 | - | 8 - 9 | 1503 | 7191761 | 137 |
| FQ223354.1 | - | 1 - 2 | 186 | 137 | ||
| M80367.1 | - | 8 - 9 | 1542 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gbp2
Genomic Map