Assay Details
| Gene Symbol: Clk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134340.1 | NP_599167.1 | 7 - 8 | 866 | 58 | |
| XM_006243146.2 | XP_006243208.1 | 7 - 8 | 1811 | 58 | ||
| XM_006243149.2 | XP_006243211.1 | 7 - 8 | 828 | 58 | ||
| XM_008766202.1 | XP_008764424.1 | 4 - 5 | 560 | 58 | ||
| GenBank mRNA | BC070891.1 | - | 7 - 8 | 842 | 7099115 | 58 |
| X94351.1 | - | 7 - 8 | 866 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clk3
Genomic Map