Assay Details
| Gene Symbol: Epcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138541.1 | NP_612550.1 | 5 - 6 | 616 | 124 | |
| GenBank mRNA | AJ001044.1 | - | 5 - 6 | 671 | 124 | |
| BC061787.1 | - | 5 - 6 | 620 | 124 | ||
| BC072691.1 | - | 5 - 6 | 640 | 7098920 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Epcam
Genomic Map