Assay Details
| Gene Symbol: Ubxn11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138853.2 | NP_620208.1 | 10 - 11 | 906 | 65 | |
| XM_006239000.1 | XP_006239062.1 | 10 - 11 | 968 | 65 | ||
| XM_006239002.1 | XP_006239064.1 | 8 - 9 | 842 | 65 | ||
| XM_006239004.1 | XP_006239066.1 | 8 - 9 | 822 | 65 | ||
| XM_006239005.2 | XP_006239067.1 | 7 - 8 | 1176 | 65 | ||
| XM_008764131.1 | XP_008762353.1 | 8 - 9 | 974 | 65 | ||
| XM_008764132.1 | XP_008762354.1 | 8 - 9 | 1015 | 65 | ||
| GenBank mRNA | AB072920.1 | - | 9 - 10 | 834 | 65 | |
| BC078730.1 | - | 10 - 11 | 906 | 7134156 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubxn11
Genomic Map