Assay Details
Gene Symbol: RGD621098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139040.1 | NP_620609.1 | 6 - 7 | 667 | 118 | |
GenBank mRNA | U92010.1 | - | 6 - 7 | 667 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
