Assay Details
| Gene Symbol: Lgals3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139096.1 | NP_620796.1 | 1 - 2 | 130 | 64 | |
| GenBank mRNA | AF065438.1 | - | 1 - 2 | 130 | 64 | |
| BC081724.1 | - | 1 - 2 | 103 | 7123606 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF065438.1, BC081724.1, NM_139096.1 transcripts.
Set Membership: |
|

Lgals3bp
Genomic Map