Assay Details
| Gene Symbol: Gpr116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139110.1 | NP_620810.1 | 13 - 14 | 2011 | 105 | |
| XM_006244594.2 | XP_006244656.1 | 14 - 15 | 2346 | 105 | ||
| XM_006244597.2 | XP_006244659.1 | 13 - 14 | 2225 | 105 | ||
| XM_008766832.1 | XP_008765054.1 | 15 - 16 | 2414 | 105 | ||
| XM_008766833.1 | XP_008765055.1 | 15 - 16 | 2339 | 105 | ||
| XM_008766834.1 | XP_008765056.1 | 15 - 16 | 2415 | 105 | ||
| XM_008766835.1 | XP_008765057.1 | 14 - 15 | 2293 | 105 | ||
| GenBank mRNA | AB019120.1 | - | 13 - 14 | 2011 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr116
Genomic Map