Assay Details
| Gene Symbol: Nradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139259.2 | NP_640352.1 | 3 - 4 | 415 | 70 | |
| XM_006243921.2 | XP_006243983.1 | 3 - 4 | 419 | 70 | ||
| GenBank mRNA | AF497263.1 | - | 3 - 4 | 419 | 70 | |
| AF497264.1 | - | 3 - 4 | 267 | 70 | ||
| AF534395.1 | - | 2 - 3 | 231 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nradd
Genomic Map