Assay Details
| Gene Symbol: Fmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144737.2 | NP_653338.2 | 3 - 4 | 414 | 109 | |
| XM_008769604.1 | XP_008767826.1 | 2 - 3 | 678 | 109 | ||
| XM_008769605.1 | XP_008767827.1 | 3 - 4 | 488 | 109 | ||
| GenBank mRNA | AF458414.1 | - | 3 - 4 | 381 | 109 | |
| BC070904.1 | - | 3 - 4 | 414 | 7100688 | 109 | |
| CK365136.1 | - | 3 - 4 | 349 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fmo2
Genomic Map