Assay Details
| Gene Symbol: Tnfsf15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145765.1 | NP_665708.1 | 1 - 2 | 218 | 79 | |
| GenBank mRNA | AF520787.1 | - | 1 - 2 | 218 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tnfsf15
Genomic Map