Assay Details
| Gene Symbol: Gpr56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152242.2 | NP_689448.2 | 9 - 10 | 1455 | 68 | |
| XM_006255066.2 | XP_006255128.1 | 9 - 10 | 2507 | 68 | ||
| XM_008772264.1 | XP_008770486.1 | 9 - 10 | 1657 | 68 | ||
| XM_008772265.1 | XP_008770487.1 | 10 - 11 | 1921 | 68 | ||
| XM_008772266.1 | XP_008770488.1 | 9 - 10 | 3167 | 68 | ||
| XM_008772267.1 | XP_008770489.1 | 9 - 10 | 3430 | 68 | ||
| XM_008772268.1 | XP_008770490.1 | 10 - 11 | 1754 | 68 | ||
| XM_008772269.1 | XP_008770491.1 | 9 - 10 | 2536 | 68 | ||
| XM_008772270.1 | XP_008770492.1 | 9 - 10 | 3012 | 68 | ||
| GenBank mRNA | AF529886.1 | - | 9 - 10 | 1456 | 68 | |
| EX488169.1 | - | 6 - 7 | 566 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpr56
Genomic Map