Assay Details
| Gene Symbol: Reg3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173097.1 | NP_775120.1 | 3 - 4 | 222 | 83 | |
| XM_008763019.1 | XP_008761241.1 | 3 - 4 | 360 | 83 | ||
| GenBank mRNA | L20869.1 | - | 3 - 4 | 222 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Reg3g
Genomic Map