Assay Details
| Gene Symbol: Ren | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012642.4 | NP_036774.4 | 6 - 7 | 826 | 64 | |
| GenBank mRNA | BC078878.1 | - | 6 - 7 | 826 | 7126969 | 64 |
| J02941.1 | - | 6 - 7 | 1019 | 64 | ||
| S60054.1 | - | 6 - 7 | 836 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ren
Genomic Map