Assay Details
| Gene Symbol: Sts | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012661.1 | NP_036793.1 | 7 - 8 | 1618 | 75 | |
| XM_006256960.2 | XP_006257022.1 | 7 - 8 | 1865 | 75 | ||
| XM_006256961.2 | XP_006257023.1 | 7 - 8 | 1853 | 75 | ||
| XM_006256962.2 | XP_006257024.1 | 7 - 8 | 1540 | 75 | ||
| GenBank mRNA | BC085747.1 | - | 4 - 5 | 2294 | 75 | |
| U37138.1 | - | 7 - 8 | 1618 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sts
Genomic Map