Assay Details
| Gene Symbol: Adrbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012776.1 | NP_036908.1 | 19 - 20 | 2041 | 77 | |
| GenBank mRNA | M87854.1 | - | 19 - 20 | 2041 | 77 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of the force of heart contraction
desensitization of G-protein coupled receptor protein signaling pathway
negative regulation of the force of heart contraction by chemical signal
protein phosphorylation
response to oxidative stress
G-protein coupled receptor signaling pathway
tachykinin receptor signaling pathway
heart development
aging
response to organic cyclic compound
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
receptor internalization
positive regulation of catecholamine secretion
termination of G-protein coupled receptor signaling pathway
response to hydrogen peroxide
follicle-stimulating hormone signaling pathway
negative regulation of G-protein coupled receptor protein signaling pathway
negative regulation of striated muscle contraction
cardiac muscle contraction
protein kinase activity
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
protein binding
ATP binding
Edg-2 lysophosphatidic acid receptor binding
beta-adrenergic receptor kinase activity
regulation of the force of heart contraction
desensitization of G-protein coupled receptor protein signaling pathway
negative regulation of the force of heart contraction by chemical signal
protein phosphorylation
response to oxidative stress
G-protein coupled receptor signaling pathway
tachykinin receptor signaling pathway
heart development
aging
response to organic cyclic compound
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
receptor internalization
positive regulation of catecholamine secretion
termination of G-protein coupled receptor signaling pathway
response to hydrogen peroxide
follicle-stimulating hormone signaling pathway
negative regulation of G-protein coupled receptor protein signaling pathway
negative regulation of striated muscle contraction
cardiac muscle contraction
protein kinase activity
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
protein binding
ATP binding
Edg-2 lysophosphatidic acid receptor binding
beta-adrenergic receptor kinase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P491621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163589 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P491621 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P491621 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P491621 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P491621 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700292421 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700292421 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10727548 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727543 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727546 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727552 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727538 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727555 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727544 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727551 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727549 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727556 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727547 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727557 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727553 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727542 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727554 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727541 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727540 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727550 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727545 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727539 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10727537 | - | - |
| Rat Genome 230 2.0 Array | 1387429_at | - | - |
| Rat Genome 230 2.0 Array | 1388757_at | - | - |
| Rat Genome 230 2.0 Array | 1388757_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387429_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387429_at | - | - |
| Affymetrix HT Rat Focus Array | 1388757_at | - | - |
| Affymetrix HT Rat Focus Array | 1387429_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388757_PM_at | - | - |
| Rat Expression Set 230 | 1387429_at | - | - |
| Rat Expression Set 230 | 1388757_at | - | - |
| Rat Expression Set 230 | 1388757_PM_at | - | - |
| Rat Expression Set 230 | 1387429_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10727537 | - | - |
| Rat Genome U34 Set | M87854_at | - | - |
| Rat Genome U34 Set | S48813_s_at | - | - |
| Rat Genome U34 Set | rc_AA998430_at | - | - |
| Rat Genome U34 Set | S48813_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21895880 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376677 | - | - |

Adrbk1
Genomic Map