Assay Details
| Gene Symbol: Pou1f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013008.3 | NP_037140.2 | 4 - 5 | 722 | 67 | |
| XM_006247974.1 | XP_006248036.1 | 4 - 5 | 1006 | 67 | ||
| GenBank mRNA | AB622208.1 | - | 3 - 4 | 733 | 67 | |
| BC061563.1 | - | 4 - 5 | 726 | 6918965 | 67 | |
| BC061564.1 | - | 4 - 5 | 741 | 67 | ||
| L01506.1 | - | 4 - 5 | 690 | 67 | ||
| L01507.1 | - | 4 - 5 | 766 | 67 | ||
| M23253.1 | - | 4 - 5 | 636 | 67 | ||
| X12658.1 | - | 4 - 5 | 659 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pou1f1
Genomic Map