Assay Details
| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016986.2 | NP_058682.2 | 5 - 6 | 493 | 121 | |
| GenBank mRNA | CK359511.1 | - | 5 - 6 | 408 | 121 | |
| J02791.1 | - | 5 - 6 | 408 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acadm
Genomic Map