Assay Details
| Gene Symbol: Adcyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016989.2 | NP_058685.1 | 2 - 3 | 402 | 63 | |
| XM_006245672.2 | XP_006245734.1 | 3 - 4 | 1098 | 63 | ||
| XM_006245673.2 | XP_006245735.1 | 3 - 4 | 1474 | 63 | ||
| XM_006245674.2 | XP_006245736.1 | 3 - 4 | 1427 | 63 | ||
| XM_008767418.1 | XP_008765640.1 | 3 - 4 | 1051 | 63 | ||
| XM_008767419.1 | XP_008765641.1 | 1 - 2 | 448 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adcyap1
Genomic Map