Assay Details
| Gene Symbol: Prl4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017036.2 | NP_058732.2 | 1 - 2 | 62 | 73 | |
| GenBank mRNA | BC097332.1 | - | 1 - 2 | 62 | 7443660 | 73 |
| FQ228460.1 | - | 1 - 2 | 97 | 73 | ||
| M13750.1 | - | 1 - 2 | 45 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prl4a1
Genomic Map