Assay Details
| Gene Symbol: Tceb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017103.2 | NP_058799.2 | 4 - 5 | 1488 | 126 | |
| GenBank mRNA | BC081815.1 | - | 4 - 5 | 1488 | 7110428 | 126 |
| L46816.1 | - | 1503 | 126 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tceb3
Genomic Map