Assay Details
| Gene Symbol: Gabrr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017292.2 | NP_058988.1 | 4 - 5 | 683 | 66 | |
| XM_008763610.1 | XP_008761832.1 | 6 - 7 | 669 | 66 | ||
| XM_008763611.1 | XP_008761833.1 | 3 - 4 | 1615 | 66 | ||
| GenBank mRNA | D38494.1 | - | 4 - 5 | 683 | 66 | |
More Information
Set Membership: |
|

Gabrr2
Genomic Map