Assay Details
| Gene Symbol: Tac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019162.2 | NP_062035.1 | 5 - 6 | 409 | 64 | |
| GenBank mRNA | FQ224503.1 | - | 2 - 3 | 151 | 64 | |
| M16410.1 | - | 5 - 6 | 409 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tac3
Genomic Map