Assay Details
| Gene Symbol: Rnf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019182.2 | NP_062055.1 | 2 - 3 | 281 | 123 | |
| XM_008770271.1 | XP_008768493.1 | 2 - 3 | 313 | 123 | ||
| XM_008770272.1 | XP_008768494.1 | 2 - 3 | 268 | 123 | ||
| XM_008770273.1 | XP_008768495.1 | 2 - 3 | 401 | 123 | ||
| GenBank mRNA | AF022081.1 | - | 2 - 3 | 199 | 123 | |
| AY050655.1 | - | 2 - 3 | 293 | 123 | ||
| BC062024.1 | - | 2 - 3 | 281 | 5623495 | 123 | |
| FQ211855.1 | - | 1 - 2 | 163 | 123 | ||
| FQ223606.1 | - | 2 - 3 | 297 | 123 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF022081.1, AY050655.1, BC062024.1, NM_019182.2, XM_008770271.1, XM_008770272.1, XM_008770273.1 transcripts.
Set Membership: |
|

Rnf4
Genomic Map