Assay Details
| Gene Symbol: Sepp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083911.1 | NP_001077380.1 | 2 - 3 | 344 | 136 | |
| NM_019192.2 | NP_062065.2 | 2 - 3 | 272 | 136 | ||
| GenBank mRNA | BC059137.1 | - | 2 - 3 | 273 | 136 | |
| BC072539.1 | - | 2 - 3 | 261 | 7104195 | 136 | |
| D25221.1 | - | 2 - 3 | 253 | 136 | ||
| DN933732.1 | - | 2 - 3 | 592 | 136 | ||
| M63574.1 | - | 232 | 136 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Sepp1
Genomic Map