Assay Details
| Gene Symbol: Cr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005265.1 | NP_001005265.1 | 4 - 5 | 721 | 120 | |
| NM_001005330.1 | NP_001005330.1 | 721 | 120 | |||
| NM_019301.2 | NP_062174.1 | 4 - 5 | 721 | 120 | ||
| GenBank mRNA | BC061736.1 | - | 4 - 5 | 490 | 5599318 | 120 |
| D42114.1 | - | 4 - 5 | 530 | 120 | ||
| D42115.1 | - | 4 - 5 | 466 | 120 | ||
| D42116.1 | - | 1 - 2 | 74 | 120 | ||
| L19118.1 | - | 1 - 2 | 84 | 120 | ||
| L36532.1 | - | 4 - 5 | 489 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the D42116.1 transcripts.
Set Membership: |
|

Cr1l
Genomic Map