Assay Details
| Gene Symbol: Tpsab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019322.2 | NP_062195.2 | 4 - 5 | 876 | 104 | |
| GenBank mRNA | BC061983.1 | - | 4 - 5 | 858 | 5598863 | 104 |
| U67910.1 | - | 4 - 5 | 875 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tpsab1
Genomic Map