Assay Details
| Gene Symbol: LOC100911725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243892.2 | XP_006243954.1 | 1186 | 78 | ||
| Gene Symbol: Pfkfb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019333.1 | NP_062206.1 | 12 - 13 | 1255 | 78 | |
| XM_006243840.2 | XP_006243902.1 | 11 - 12 | 1214 | 78 | ||
| XM_006243841.2 | XP_006243903.1 | 11 - 12 | 1208 | 78 | ||
| XM_006243842.2 | XP_006243904.1 | 11 - 12 | 1176 | 78 | ||
| XM_008766563.1 | XP_008764785.1 | 11 - 12 | 2255 | 78 | ||
| GenBank mRNA | EU404101.1 | - | 11 - 12 | 1202 | 78 | |
| M64797.1 | - | 12 - 13 | 1255 | 78 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911725
Genomic Map