Assay Details
| Gene Symbol: LOC100911725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006243892.2 | XP_006243954.1 | 808 | 80 | ||
| Gene Symbol: Pfkfb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019333.1 | NP_062206.1 | 9 - 10 | 877 | 80 | |
| XM_006243840.2 | XP_006243902.1 | 8 - 9 | 836 | 80 | ||
| XM_006243841.2 | XP_006243903.1 | 8 - 9 | 830 | 80 | ||
| XM_006243842.2 | XP_006243904.1 | 8 - 9 | 798 | 80 | ||
| XM_008766563.1 | XP_008764785.1 | 8 - 9 | 1877 | 80 | ||
| GenBank mRNA | EU404101.1 | - | 8 - 9 | 846 | 80 | |
| EU404102.1 | - | 8 - 9 | 846 | 80 | ||
| EU404103.1 | - | 8 - 9 | 846 | 80 | ||
| M64797.1 | - | 9 - 10 | 877 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911725
Genomic Map