Assay Details
| Gene Symbol: Cnn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019359.1 | NP_062232.1 | 5 - 6 | 736 | 57 | |
| GenBank mRNA | BC062020.1 | - | 5 - 6 | 739 | 5623281 | 57 |
| FQ228936.1 | - | 2 - 3 | 173 | 57 | ||
| U06755.1 | - | 5 - 6 | 736 | 57 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cnn3
Genomic Map