Assay Details
| Gene Symbol: LOC102549615 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006252223.2 | XP_006252285.1 | 1 - 2 | 246 | 110 | |
| Gene Symbol: Extl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020097.1 | NP_064482.1 | 3 - 4 | 2427 | 110 | |
| XM_006252206.2 | XP_006252268.1 | 6 - 7 | 3733 | 110 | ||
| GenBank mRNA | AB033367.1 | - | 3 - 4 | 2427 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC102549615
Genomic Map