Assay Details
| Gene Symbol: Prl8a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022846.2 | NP_074037.1 | 4 - 5 | 354 | 86 | |
| XM_006253906.1 | XP_006253968.1 | 4 - 5 | 414 | 86 | ||
| GenBank mRNA | BC097316.1 | - | 4 - 5 | 355 | 7442457 | 86 |
| FQ219955.1 | - | 4 - 5 | 414 | 86 | ||
| FQ220067.1 | - | 4 - 5 | 413 | 86 | ||
| FQ222955.1 | - | 4 - 5 | 414 | 86 | ||
| FQ229664.1 | - | 4 - 5 | 388 | 86 | ||
| FQ229702.1 | - | 4 - 5 | 412 | 86 | ||
| FQ229882.1 | - | 4 - 5 | 414 | 86 | ||
| L06441.1 | - | 4 - 5 | 356 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prl8a2
Genomic Map