Assay Details
| Gene Symbol: Necab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133415.1 | NP_596906.1 | 8 - 9 | 758 | 60 | |
| XM_006255703.2 | XP_006255765.1 | 6 - 7 | 1824 | 60 | ||
| GenBank mRNA | AF193757.1 | - | 8 - 9 | 758 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Necab2
Genomic Map