Assay Details
| Gene Symbol: Caly | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190399.1 | NP_001177328.1 | 3 - 4 | 358 | 67 | |
| NM_138915.1 | NP_620270.1 | 3 - 4 | 417 | 67 | ||
| GenBank mRNA | AF303658.1 | - | 3 - 4 | 417 | 67 | |
| FQ213422.1 | - | 3 - 4 | 332 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Caly
Genomic Map