Assay Details
| Gene Symbol: Klra22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173291.1 | NP_775413.1 | 2 - 3 | 251 | 68 | |
| GenBank mRNA | AY649838.1 | - | 2 - 3 | 262 | 68 | |
| U56822.1 | - | 2 - 3 | 251 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Klra22
Genomic Map