Assay Details
| Gene Symbol: Ntf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013184.3 | NP_037316.2 | 2 - 3 | 192 | 121 | |
| GenBank mRNA | S69323.1 | - | 2 - 3 | 192 | 121 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_013184.3, S69323.1 transcripts.
Set Membership: |
|

Ntf4
Genomic Map