Assay Details
| Gene Symbol: Trpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083115.1 | NP_001076584.1 | 2 - 3 | 902 | 83 | |
| NM_080396.1 | NP_536321.1 | 2 - 3 | 902 | 83 | ||
| XM_006232356.2 | XP_006232418.1 | 3 - 4 | 1226 | 83 | ||
| XM_008761008.1 | XP_008759230.1 | 3 - 4 | 1226 | 83 | ||
| GenBank mRNA | AB008889.2 | - | 3 - 4 | 1016 | 83 | |
| AF288407.1 | - | 2 - 3 | 918 | 83 | ||
| AF288408.1 | - | 2 - 3 | 918 | 83 | ||
| AF421363.1 | - | 2 - 3 | 902 | 83 | ||
| AF421364.1 | - | 2 - 3 | 902 | 83 | ||
| AF421365.1 | - | 2 - 3 | 902 | 83 | ||
| AF421366.1 | - | 2 - 3 | 902 | 83 | ||
| AF421367.1 | - | 2 - 3 | 902 | 83 | ||
| AF421368.1 | - | 2 - 3 | 902 | 83 | ||
| FQ228645.1 | - | 3 - 4 | 1121 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trpc4
Genomic Map