Assay Details
| Gene Symbol: Cnga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053496.2 | NP_445948.2 | 2 - 3 | 273 | 70 | |
| GenBank mRNA | U12425.1 | - | 2 - 3 | 272 | 70 | |
| U12623.1 | - | 2 - 3 | 274 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnga4
Genomic Map