Assay Details
| Gene Symbol: Rdh7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133543.3 | NP_598227.3 | 659 | 88 | ||
| GenBank mRNA | BC088090.1 | - | 659 | 7368214 | 88 | |
| BC098650.1 | - | 721 | 7371059 | 88 | ||
| FQ209576.1 | - | 643 | 88 | |||
| U18762.1 | - | 868 | 88 | |||
| U33501.1 | - | 640 | 88 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Rdh7
Genomic Map