Assay Details
| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145676.1 | NP_663709.1 | 3 - 4 | 267 | 124 | |
| XM_006241967.2 | XP_006242029.1 | 4 - 5 | 384 | 124 | ||
| GenBank mRNA | AF393211.1 | - | 3 - 4 | 267 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cby1
Genomic Map