Assay Details
| Gene Symbol: Igf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082477.2 | NP_001075946.2 | 2 - 3 | 204 | 69 | |
| NM_001082478.1 | NP_001075947.1 | 2 - 3 | 501 | 69 | ||
| NM_001082479.1 | NP_001075948.1 | 2 - 3 | 501 | 69 | ||
| NM_178866.4 | NP_849197.3 | 2 - 3 | 204 | 69 | ||
| XM_006241191.2 | XP_006241253.1 | 3 - 4 | 345 | 69 | ||
| XM_006241192.1 | XP_006241254.1 | 3 - 4 | 259 | 69 | ||
| XM_006241193.2 | XP_006241255.1 | 3 - 4 | 344 | 69 | ||
| XM_006241194.2 | XP_006241256.1 | 2 - 3 | 383 | 69 | ||
| GenBank mRNA | BC086374.1 | - | 2 - 3 | 204 | 7300903 | 69 |
| D00698.1 | - | 2 - 3 | 185 | 69 | ||
| FQ209824.1 | - | 3 - 4 | 333 | 69 | ||
| FQ210560.1 | - | 2 - 3 | 217 | 69 | ||
| FQ210688.1 | - | 2 - 3 | 217 | 69 | ||
| FQ220385.1 | - | 2 - 3 | 525 | 69 | ||
| FQ225388.1 | - | 2 - 3 | 217 | 69 | ||
| FQ225437.1 | - | 2 - 3 | 217 | 69 | ||
| FQ231951.1 | - | 2 - 3 | 217 | 69 | ||
| M15480.1 | - | 2 - 3 | 248 | 69 | ||
| M15481.1 | - | 2 - 3 | 936 | 69 | ||
| X06043.1 | - | 1 - 2 | 86 | 69 | ||
| X06107.1 | - | 2 - 3 | 479 | 69 | ||
| X06108.1 | - | 3 - 4 | 293 | 69 | ||
More Information
Set Membership: |
|

Igf1
Genomic Map